missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£148.00 - £294.00
Specifications
| Antigen | SOCS-5 |
|---|---|
| Dilution | Western Blot 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18009113
|
Novus Biologicals
NB100-56639SS |
0.05 mL |
£148.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18767133
|
Novus Biologicals
NB100-56639 |
0.2 mL |
£294.00
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOCS-5 Polyclonal specifically detects SOCS-5 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| SOCS-5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS containing 0.1 mg/ml BSA. with 0.02% Sodium Azide | |
| CIS6Cytokine-inducible SH2 protein 6, Cish5, CISH6CIS-6, KIAA0671Cytokine-inducible SH2-containing protein 5, SOCS-5CISH5, suppressor of cytokine signaling 5 | |
| SOCS5 | |
| IgG | |
| Protein G purified |
| Western Blot 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| O75159 | |
| 9655 | |
| This polyclonal antibody was raised against a synthetic peptide containing amino acid residues of human SOCS5 protein. Human or mouse spleen lysate can be used as a positive control. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title