missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56639
This item is not returnable.
View return policy
Description
SOCS-5 Polyclonal specifically detects SOCS-5 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| SOCS-5 | |
| Polyclonal | |
| Western Blot 1:500 | |
| O75159 | |
| SOCS5 | |
| This polyclonal antibody was raised against a synthetic peptide containing amino acid residues of human SOCS5 protein. Human or mouse spleen lysate can be used as a positive control. | |
| 0.2 mL | |
| Signal Transduction | |
| 9655 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS containing 0.1 mg/ml BSA. with 0.02% Sodium Azide | |
| CIS6Cytokine-inducible SH2 protein 6, Cish5, CISH6CIS-6, KIAA0671Cytokine-inducible SH2-containing protein 5, SOCS-5CISH5, suppressor of cytokine signaling 5 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction