missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £310.00
Specifications
| Antigen | SOCS-3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 5ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18156452
|
Novus Biologicals
NBP2-27116SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18109133
|
Novus Biologicals
NBP2-27116 |
0.1 mg |
£310.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOCS-3 Polyclonal specifically detects SOCS-3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SOCS-3 | |
| Immunohistochemistry-Paraffin 5ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3 | |
| SOCS3 | |
| IgG | |
| Protein A purified | |
| The SOCS3 amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, cow, guinea pig, dog, Chinese hamster, horse, mole rat, elephant, rhesus monkey, sheep, pig, and pole-cat, 94% in panda, Carolina anole, new world monkey, chicken, wild turkey, opossum, and zebra finch, 89% in platypus, 72% in channel catfish, and 67% homologous in frog. |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 9021 | |
| A portion of amino acids 170-220 from human SOCS3 was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title