missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27116SS
This item is not returnable.
View return policy
Description
SOCS-3 Polyclonal specifically detects SOCS-3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SOCS-3 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| SOCS3 | |
| A portion of amino acids 170-220 from human SOCS3 was used as the immunogen for this antibody. | |
| 0.025 mg | |
| Signal Transduction | |
| 9021 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Immunohistochemistry-Paraffin 5ug/ml | |
| CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The SOCS3 amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, cow, guinea pig, dog, Chinese hamster, horse, mole rat, elephant, rhesus monkey, sheep, pig, and pole-cat, 94% in panda, Carolina anole, new world monkey, chicken, wild turkey, opossum, and zebra finch, 89% in platypus, 72% in channel catfish, and 67% homologous in frog. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction