missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX29 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79261
This item is not returnable.
View return policy
Description
SNX29 Polyclonal specifically detects SNX29 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SNX29 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| A-388D4.1, FLJ00143, FLJ12363, FLJ40609, RUN domain containing 2A, sorting nexin 29 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 92017 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_115543 | |
| SNX29 | |
| Synthetic peptide directed towards the N terminal of human RUNDC2AThe immunogen for this antibody is RUNDC2A. Peptide sequence MSGSQNNDKRQFLLERLLDAVKQCQIRFGGRKEIASDSDSRVTCLCAQFE. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Human: 100%; Green puffer: 92%; Black-legged tick: 78%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Rabbit, Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction