missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £302.00
Specifications
| Antigen | SNX2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.25-0.50 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18116185
|
Novus Biologicals
NBP2-24514SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18167841
|
Novus Biologicals
NBP2-24514 |
0.1 mg |
£302.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SNX2 Polyclonal specifically detects SNX2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot.Specifications
| SNX2 | |
| Western Blot 0.25-0.50 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 6643 | |
| A portion of amino acids 120-170 of human Snx2 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Primate | |
| MGC5204, sorting nexin 2, sorting nexin-2, Transformation-related gene 9 protein, TRG-9 | |
| SNX2 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse and bovine, and 77% homologous in chicken. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title