missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24514SS
This item is not returnable.
View return policy
Description
SNX2 Polyclonal specifically detects SNX2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot.
Specifications
| SNX2 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| SNX2 | |
| A portion of amino acids 120-170 of human Snx2 was used as the immunogen. | |
| 0.025 mg | |
| Signal Transduction | |
| 6643 | |
| Human, Mouse, Rat, Primate | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.25-0.50 ug/ml | |
| MGC5204, sorting nexin 2, sorting nexin-2, Transformation-related gene 9 protein, TRG-9 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse and bovine, and 77% homologous in chicken. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction