missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38045
This item is not returnable.
View return policy
Description
SNRPG Polyclonal specifically detects SNRPG in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SNRPG | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| P62308 | |
| SNRPG | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RHVQGILRGFDPFMNLVIDECVEMATSGQQ | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 6637 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MGC117317, PBSCG, small nuclear ribonucleoprotein G, small nuclear ribonucleoprotein polypeptide G, SMG, Sm-GSm protein G, snRNP-G | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido