missing translation for 'onlineSavingsMsg'
Learn More

SNAP43 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£378.00

Specifications

Antigen SNAP43
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18277675
View Documents
Novus Biologicals
NBP1-80296
100 μL
£378.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SNAP43 Polyclonal specifically detects SNAP43 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SNAP43
Polyclonal
Purified
RUO
NP_003073
6617
Synthetic peptide directed towards the C terminal of human SNAPC1. Peptide sequence GQGQVKATRKKEKKERLKPAGRKMSLRNKGNVQNIHKEDKPLSLSMPVIT.
Primary
Western Blot
Unconjugated
Rabbit
Human
Proximal sequence element-binding transcription factor subunit gamma, PSE-binding factor subunit gamma, PTF subunit gamma, PTFgamma, small nuclear RNA activating complex, polypeptide 1, 43kD, small nuclear RNA activating complex, polypeptide 1, 43kDa, Small nuclear RNA-activating complex polypeptide 1, SNAP43SNAPc 43 kDa subunit, SNAPc subunit 1, snRNA-activating protein complex 43 kDa subunit, snRNA-activating protein complex subunit 1
SNAPC1
IgG
Protein A purified
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.