missing translation for 'onlineSavingsMsg'
Learn More

SNAP25, Rabbit anti-Human,Mouse,Rat,Porcine,Bovine,Feline,Guinea Pig,Hamster, Clone: SP12, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP2-81057-0.025MG

 View more versions of this product

Product Code. 18230549

  • £217.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SNAP25 Monoclonal specifically detects SNAP25 in Human, Mouse, Rat, Porcine, Bovine, Feline, Guinea Pig, Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SNAP25
Monoclonal
1 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa
Rabbit
Protein A purified
RUO
Primary
This antibody binds to native and denatured SNAP25.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG κ
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
SP12
Unconjugated
PBS
SNAP25
Recombinant monoclonal antibody to SNAP25.
0.025 mg
Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
6616
Human, Mouse, Rat, Pig, Bovine, Feline, Guinea Pig, Hamster
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.