missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAP25, Rabbit anti-Human,Mouse,Rat,Porcine,Bovine,Feline,Guinea Pig,Hamster, Clone: SP12, Novus Biologicals™
Rabbit Monoclonal Antibody
£217.00 - £463.00
Specifications
| Antigen | SNAP25 |
|---|---|
| Clone | SP12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18230549
|
Novus Biologicals
NBP2-81057-0.025MG |
0.025 mg |
£217.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18314670
|
Novus Biologicals
NBP2-81057-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SNAP25 Monoclonal specifically detects SNAP25 in Human, Mouse, Rat, Porcine, Bovine, Feline, Guinea Pig, Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| SNAP25 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| PBS | |
| 6616 | |
| Recombinant monoclonal antibody to SNAP25. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| SP12 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Feline, Guinea Pig, Hamster | |
| bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa | |
| SNAP25 | |
| IgG κ | |
| Protein A purified | |
| This antibody binds to native and denatured SNAP25. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title