Learn More
Macherey-Nagel™ Snap Tubes
Conical centrifuge tubes with snap lids for use as lysis tubes for incubation of samples in the purification of nucleic acids with compatible kits (e.g. NucleoSnap Plasmid Midi or NucleoMag DNA Plasma). Packs of 50 tubes.
Brand: Macherey-Nagel™ 740822.50
Additional Details : Weight : 0.75000kg
Specifications
Clear | |
Centrifuge | |
Snap lid | |
50 Pack |
Conical Tube | |
Incubation and lysis of samples | |
Centrifuge Tube | |
50 mL |