missing translation for 'onlineSavingsMsg'
Learn More

Snail, p Ser246 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-54776-0.05MG

 View more versions of this product

Product Code. 18204854

  • £346.00 / 0.05mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Snail Polyclonal specifically detects Snail in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Snail
Polyclonal
Unconjugated
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl and 50% glycerol. with 0.02% sodium azide
SNAI1
SNAI1 (phospho Ser246) antibody was raised against a peptide sequence around phosphorylation site of Serine 246 (T-F-S (p)-R-M) derived from Human SNAI1.
Primary
The antibody detects endogenous levels of SNAI1 only when phosphorylated at serine 246.
Store at -20°C. Avoid freeze/thaw cycles.
Western Blot, Immunofluorescence
1 mg/mL
Western Blot 1:500-1:1000, Immunofluorescence 1:100-1:200
Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAHdJ710H13.1, snail 1 (drosophila homolog), zinc finger protein, snail 1 homolog, snail 1 zinc finger protein, snail 1, zinc finger protein, snail homolog 1 (Drosophila), zinc finger protein SNAI1
Rabbit
0.05 mg
6615
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only