missing translation for 'onlineSavingsMsg'
Learn More

Snail, p Ser246 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£163.00 - £539.00

Specifications

Concentration 1 mg/mL
Dilution Western Blot 1:500-1:1000, Immunofluorescence 1:100-1:200
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18251131
View Documents
Novus Biologicals
NBP2-54776
0.1 mg
£539.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18204854
View Documents
Novus Biologicals
NBP2-54776-0.05MG
0.05 mg
£346.00
0.05mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18657486
View Documents
Novus Biologicals
NBP2-54776-0.025mg
0.025 mg
£163.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Snail Polyclonal specifically detects Snail in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence.
Specifications

Specifications

1 mg/mL
Western Blot, Immunofluorescence
Unconjugated
Human, Mouse
6615
SNAI1 (phospho Ser246) antibody was raised against a peptide sequence around phosphorylation site of Serine 246 (T-F-S (p)-R-M) derived from Human SNAI1.
Primary
Western Blot 1:500-1:1000, Immunofluorescence 1:100-1:200
Polyclonal
Rabbit
Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAHdJ710H13.1, snail 1 (drosophila homolog), zinc finger protein, snail 1 homolog, snail 1 zinc finger protein, snail 1, zinc finger protein, snail homolog 1 (Drosophila), zinc finger protein SNAI1
SNAI1
IgG
The antibody detects endogenous levels of SNAI1 only when phosphorylated at serine 246.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.