missing translation for 'onlineSavingsMsg'
Learn More

SN-38, TRC

Product Code. 30004004
Change view
Click to view available options
Quantity:
5 mg
50 mg
250 mg
Unit Size:
250mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004004 50 mg 50mg
30007318 5 mg 5mg
30010980 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004004 Supplier Toronto Research Chemicals Supplier No. TRCS58995050MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86639-52-3
Molecular Formula C22 H20 N2 O5
Molecular Weight (g/mol) 392.4
Synonym 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 4,11-diethyl-4,9-dihydroxy-, (4S)-, (S)-4,11-Diethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 10-Hydroxy-7-ethylcamptothecin, 7-Ethyl-10-hydroxy-20(S)-camptothecin, 7-Ethyl-10-hydroxycamptothecin, (4S)-4,11-Diethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 11-Ethyl-9-hydroxycamptothecin, SN-38
IUPAC Name (S)-4,11-Diethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione
SMILES CCc1c2CN3C(=O)C4=C(C=C3c2nc5ccc(O)cc15)[C@@](O)(CC)C(=O)OC4

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H20N2O5/c1-3-12-13-7-11(25)5-6-17(13)23-19-14(12)9-24-18(19)8-16-15(20(24)26)10-29-21(27)22(16,28)4-2/h5-8,25,28H,3-4,9-10H2,1-2H3/t22-/m0/s1
Formula Weight 392.1372
Percent Purity >95%
Chemical Name or Material 7-Ethyl-10-hydroxycamptothecin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.