missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMCR7/MID49 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£111.18 - £274.38
Specifications
| Antigen | SMCR7/MID49 |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
This antibody recognizes isoform1 49 kDa only.
This gene encodes an outer mitochondrial membrane protein that functions in the regulation of mitochondrial morphology. It can directly recruit the fission mediator dynamin-related protein 1 (Drp1) to the mitochondrial surface. The gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| SMCR7/MID49 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q5NCS9, Q96C03 | |
| 125170, 237781 | |
| SMCR7/MID49 Fusion Protein Ag9531 | |
| Primary | |
| -20°C | |
| MIEF2 |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| MID49, MIEF2, SMCR7 | |
| Mief2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title