missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMCR7/MID49 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16413-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody recognizes isoform1 49 kDa only.
This gene encodes an outer mitochondrial membrane protein that functions in the regulation of mitochondrial morphology. It can directly recruit the fission mediator dynamin-related protein 1 (Drp1) to the mitochondrial surface. The gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| SMCR7/MID49 | |
| Polyclonal | |
| Unconjugated | |
| MIEF2 | |
| MID49, MIEF2, SMCR7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 125170, 237781 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5NCS9, Q96C03 | |
| Mief2 | |
| SMCR7/MID49 Fusion Protein Ag9531 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction