missing translation for 'onlineSavingsMsg'
Learn More

SMARCD3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen SMARCD3
Dilution Western Blot 1:500 - 1:1000, ELISA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30229097
View Documents
Novus Biologicals
NBP3-35918-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30229098
View Documents
Novus Biologicals
NBP3-35918-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SMARCD3 Polyclonal antibody specifically detects SMARCD3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications

Specifications

SMARCD3
ELISA, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
60 kDa BRG-1/Brm-associated factor subunit C, 60kDa BRG-1/Brm associated factor subunit c, BAF60Cchromatin remodeling complex BAF60C subunit, BRG1-associated factor 60C, CRACD3, mammalian chromatin remodeling complex BRG1-associated factor 60C, MGC111010, Rsc6p, subfamily d, member 3, SWI/SNF complex 60 kDa subunit C, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily D member 3, Swp73-like protein
Recombinant fusion protein containing a sequence corresponding to amino acids 275-440 of human SMARCD3 (NP_001003801.1).,, Sequence:, LHTQSRSAIVQALWQYVKTNRLQDSHDKEYINGDKYFQQIFDCPRLKFSEIPQRLTALLLPPDPIVINHVISVDPSDQKKTACYDIDVEVEEPLKGQMSSFLLSTANQQEISALDSKIHETIESINQLKIQRDFMLSFSRDPKGYVQDLLRSQSRDLKVMTDVAGN
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500 - 1:1000, ELISA
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
6604
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.