missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35918-100ul
This item is not returnable.
View return policy
Description
SMARCD3 Polyclonal antibody specifically detects SMARCD3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| SMARCD3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| 60 kDa BRG-1/Brm-associated factor subunit C, 60kDa BRG-1/Brm associated factor subunit c, BAF60Cchromatin remodeling complex BAF60C subunit, BRG1-associated factor 60C, CRACD3, mammalian chromatin remodeling complex BRG1-associated factor 60C, MGC111010, Rsc6p, subfamily d, member 3, SWI/SNF complex 60 kDa subunit C, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily D member 3, Swp73-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 275-440 of human SMARCD3 (NP_001003801.1).,, Sequence:, LHTQSRSAIVQALWQYVKTNRLQDSHDKEYINGDKYFQQIFDCPRLKFSEIPQRLTALLLPPDPIVINHVISVDPSDQKKTACYDIDVEVEEPLKGQMSSFLLSTANQQEISALDSKIHETIESINQLKIQRDFMLSFSRDPKGYVQDLLRSQSRDLKVMTDVAGN | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6604 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction