Learn More
VACUUBRAND™ Small KF Flange For VARIO-SP™ Diaphragm Pumps MZ 4 and ME 4
Shop All Vacuubrand Products
Click to view available options
For Use With:
MZ/ME 4 VARIO-SP Diaphragm Pumps
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Inside | 16 mm |
| For Use With | MZ/ME 4 VARIO-SP Diaphragm Pumps |
| Quantity | 1 Each |
| Size | Small |
| Type | Small KF Flange |