missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad5 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | Smad5 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229481
|
Novus Biologicals
NBP3-33193-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30226667
|
Novus Biologicals
NBP3-33193-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Smad5 Monoclonal antibody specifically detects Smad5 in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western BlotSpecifications
| Smad5 | |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 4090 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| hSmad5, JV5-1DKFZp781O1323, MAD homolog 5, MAD, mothers against decapentaplegic homolog 5 (Drosophila), MADH5mothers against decapentaplegic homolog 5, mothers against decapentaplegic homolog 5, mothers against decapentaplegic, drosophila, homolog of, 5, Mothers against DPP homolog 5, SMA- and MAD-related protein 5, SMAD 5, SMAD family member 5DKFZp781C1895, SMAD, mothers against DPP homolog 5, SMAD, mothers against DPP homolog 5 (Drosophila), Smad5 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human Smad5 (Q99717).,, Sequence:, STYPNSPASSGPGSPFQLPADTPPPAYMPPDDQMGQDNSQPMDTSNNMIPQIMPSISSRDVQPVAYEEPKHWCSIVYYELNNRVGEAFHASSTSVLVDGFT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title