missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad2, p Ser467 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£154.00 - £491.00
Specifications
| Concentration | 1 mg/mL |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18292954
|
Novus Biologicals
NBP2-54771-0.05MG |
0.05 mg |
£335.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18220591
|
Novus Biologicals
NBP2-54771 |
0.1 mg |
£491.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666758
|
Novus Biologicals
NBP2-54771-0.025mg |
0.025 mg |
£154.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Smad2 Polyclonal specifically detects Smad2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Human, Mouse, Rat | |
| 4087 | |
| Smad2 (Phospho-Ser467) antibody was raised against a peptide sequence around phosphorylation site of serine 467 (C-S-S-M-S (p) ) derived from Human Smad2. | |
| Primary |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, pSMAD, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2 | |
| SMAD2 | |
| IgG | |
| This antibody detects endogenous level of Smad2 only when phosphorylated at serine 467. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title