missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad2, p Ser467 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54771
This item is not returnable.
View return policy
Description
Smad2 Polyclonal specifically detects Smad2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Smad2 (p Ser467) | |
| Polyclonal | |
| Unconjugated | |
| Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl and 50% glycerol with 0.02% Sodium Azide | |
| SMAD2 | |
| Smad2 (Phospho-Ser467) antibody was raised against a peptide sequence around phosphorylation site of serine 467 (C-S-S-M-S (p) ) derived from Human Smad2. | |
| Epitope affinity purified | |
| RUO | |
| Primary | |
| This antibody detects endogenous level of Smad2 only when phosphorylated at serine 467. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000 | |
| hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, pSMAD, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2 | |
| Rabbit | |
| 60 kDa | |
| 0.1 mg | |
| Cancer, Cell Cycle and Replication, Metastasis, Wnt Signaling Pathway | |
| 4087 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction