missing translation for 'onlineSavingsMsg'
Learn More

sm (Smith Antigen) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ sm (Smith Antigen) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-450-R7

 View more versions of this product

Product Code. 12651467

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

The sm (Smith) is an autoantigen on a series of particles composed of RNA and protein. Unlike RNP, the sm antigen is resistant to RNase, but partially sensitive to trypsin. It is involved in the pathogenesis of Systemic lupus erythematosus (SLE). Antibodies to sm are found in 20-30% of patients with SLE.

Host Species: Mouse

Clone: Y12

Isotype: IgG3/κ

Species Reactivity: Human, Pig, Rabbit, Rat and Drosophila. Others not known.

Epitope: Not determined

Molecular Weight: ∽25kDa (predominant)

Positive Control: Ls174T cells, Tonsil

Cellular Localization: Nuclear Ab-1 recognizes cross-reactive epitopes on the B'/B and D polypeptides of Sm. Ab-1 precipitates the small nuclear RNAs U1, U2, U4, U5, and U6 providing direct evidence that the Sm antigen resides on all these RNA-protein complexes.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Affinity Purification
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

sm (Smith Antigen) Ab-1
Monoclonal
Prediluted
Mouse
RUO
Primary
Prediluted
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Y12
Unconjugated
7 mL
DNA Repair
Drosophila, Human, Pig, Rabbit, Rat
IgG3 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.