missing translation for 'onlineSavingsMsg'
Learn More

sm (Smith Antigen) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ sm (Smith Antigen) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

£118.32 - £540.60

Specifications

Antigen sm (Smith Antigen) Ab-1
Clone Y12
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Mouse
View More Specs

Products 8
Product Code Brand Quantity Conjugate Form Concentration Price Quantity & Availability  
Product Code Brand Quantity Conjugate Form Concentration Price Quantity & Availability  
12661457
View Documents
Epredia
MS-450-B0
100 μL Biotin Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12651457
View Documents
Epredia
MS-450-B1
500 μL Biotin Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12681457
View Documents
Epredia
MS-450-P0
100 μL Unconjugated Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12611467
View Documents
Epredia
MS-450-P1ABX
100 μL Unconjugated Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12601467
View Documents
Epredia
MS-450-PABX
200 μL Unconjugated Concentrated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12691457
View Documents
Epredia
MS-450-P1
500 μL Unconjugated Concentrated 200μg/mL
ONLINE EXCLUSIVE PRICE
£386.00
£362.81 / 500µL
Save £23.19
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12671457
View Documents
Epredia
MS-450-P
1 mL Unconjugated Concentrated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12651467
View Documents
Epredia
MS-450-R7
7 mL Unconjugated Prediluted Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

The sm (Smith) is an autoantigen on a series of particles composed of RNA and protein. Unlike RNP, the sm antigen is resistant to RNase, but partially sensitive to trypsin. It is involved in the pathogenesis of Systemic lupus erythematosus (SLE). Antibodies to sm are found in 20-30% of patients with SLE.

Host Species: Mouse

Clone: Y12

Isotype: IgG3/κ

Species Reactivity: Human, Pig, Rabbit, Rat and Drosophila. Others not known.

Epitope: Not determined

Molecular Weight: ∽25kDa (predominant)

Positive Control: Ls174T cells, Tonsil

Cellular Localization: Nuclear Ab-1 recognizes cross-reactive epitopes on the B'/B and D polypeptides of Sm. Ab-1 precipitates the small nuclear RNAs U1, U2, U4, U5, and U6 providing direct evidence that the Sm antigen resides on all these RNA-protein complexes.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Affinity Purification
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

sm (Smith Antigen) Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Mouse
DNA Repair
IgG3 κ
Y12
Monoclonal
RUO
Drosophila, Human, Pig, Rabbit, Rat
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.