Learn More
Edmund Bühler GmbH™ Sliding Plate with Combifix SM
Affix to a multistory platform. Edmund Bühler GmbH™ Sliding Plate with Combifix SM fastens horizontal separating funnels of different sizes to platform.
£540.39 - £663.44
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
10540102
|
Edmund Bühler GmbH™
0051 484 |
5 Clamping Strips H, additional base plate |
£540.39
Each |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10071051
|
Edmund Bühler GmbH™
0051 485 |
4 Clamping Strips H, 4 Clamping Strips V, additional base plate |
£637.62
Each |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10019280
|
Edmund Bühler GmbH™
0051 486 |
2 Clamping Strips H, 2 Cramp Strips, 1 Spring Strip, additional base plate |
£663.44
Each |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
Description
- Fastens horizontal separating funnels of different sizes to platform
- For use with Combifix VKS or SM Universal Shakers
Specifications
Sliding Plate |