Learn More
Edmund Bühler GmbH™ Sliding Plate with Combifix SM
Affix to a multistory platform. Edmund Bühler GmbH™ Sliding Plate with Combifix SM fastens horizontal separating funnels of different sizes to platform.
Brand:  Edmund Bühler GmbH™ 0051 484
 View more versions of this product
Product Code. 10540102
  • £540.39 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fastens horizontal separating funnels of different sizes to platform
  • For use with Combifix VKS or SM Universal Shakers
Order Info
Size
Specifications
Specifications
Sliding Plate
Combifix SM
5 Clamping Strips H, additional base plate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products