Learn More
Cleaver Scientific Slide Chilling Plate
Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Comet Assay
Unit Size:
Each
Description
- Allows for rapid solidification of the low melting point agarose on the Comet Assay slide
- Facilitates easy retrieval of the slides once the agarose gels are solid
Specifications
Specifications
| Capacity | 26 Slides |
| For Use With (Equipment) | Comet Assay |
| Type | Slide Chilling Plate |
| Model | CSL-CHILLPLATE |