missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLFN5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£177.00 - £386.00
Specifications
| Antigen | SLFN5 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17703004
|
Invitrogen™
PA5121602 |
20 μL |
£177.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17792994
![]() |
Invitrogen™
PA5121601 |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLFN5 may have a role in hematopoietic cell differentiation.Specifications
| SLFN5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q08AF3 | |
| 162394 | |
| Asp658-Val891 with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SLFN5 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Schlafen family member 5; SLFN5 | |
| SLFN5 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title