missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLFN5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5121602

 View more versions of this product

Product Code. 17703004

  • £177.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SLFN5 may have a role in hematopoietic cell differentiation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLFN5
Polyclonal
Unconjugated
SLFN5
Schlafen family member 5; SLFN5
Rabbit
Antigen affinity chromatography, Protein A
RUO
162394
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q08AF3
SLFN5
Asp658-Val891 with N-terminal His Tag.
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.