missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLCO1C1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | SLCO1C1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SLCO1C1 Polyclonal specifically detects SLCO1C1 in Human samples. It is validated for Western Blot.Specifications
| SLCO1C1 | |
| Polyclonal | |
| Rabbit | |
| Q9NYB5 | |
| 53919 | |
| IgG | |
| 79 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| OATP1, OATP14Thyroxine transporter, OATP1C1Solute carrier family 21 member 14, OATPFOAT-RP-5, OATP-FOrganic anion-transporting polypeptide 14, OATPRP5, Organic anion transporter F, Organic anion transporter polypeptide-related protein 5, organic anion transporting polypeptide 14, SLC21A14OATP-14, solute carrier family 21 (organic anion transporter), member 14, solute carrier organic anion transporter family member 1C1, solute carrier organic anion transporter family, member 1C1 | |
| SLCO1C1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title