missing translation for 'onlineSavingsMsg'
Learn More

SLCO1C1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-59642

 View more versions of this product

Product Code. 18286241

  • £362.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SLCO1C1 Polyclonal specifically detects SLCO1C1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLCO1C1
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
OATP1, OATP14Thyroxine transporter, OATP1C1Solute carrier family 21 member 14, OATPFOAT-RP-5, OATP-FOrganic anion-transporting polypeptide 14, OATPRP5, Organic anion transporter F, Organic anion transporter polypeptide-related protein 5, organic anion transporting polypeptide 14, SLC21A14OATP-14, solute carrier family 21 (organic anion transporter), member 14, solute carrier organic anion transporter family member 1C1, solute carrier organic anion transporter family, member 1C1
Rabbit
79 kDa
100 μL
Primary
Yeast 83%.
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast
IgG
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
Q9NYB5
SLCO1C1
Synthetic peptides corresponding to SLCO1C1 (solute carrier organic anion transporter family, member 1C1). The peptide sequence was selected from the N terminal of SLCO1C1. Peptide sequence VDTSSSMWIYVFLGNLLRGIGETPIQPLGIAYLDDFASEDNAAFYIGCVQ.
Affinity purified
RUO
53919
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.