missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC7A7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | SLC7A7 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SLC7A7 Polyclonal specifically detects SLC7A7 in Human samples. It is validated for Western Blot.Specifications
| SLC7A7 | |
| Polyclonal | |
| Rabbit | |
| Q9UM01 | |
| 9056 | |
| IgG | |
| 56 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| LAT3, LPI, Monocyte amino acid permease 2, MOP-2, solute carrier family 7 (cationic amino acid transporter, y+ system), member 7, Solute carrier family 7 member 7, Y+L amino acid transporter 1, Y+LAT1, y+LAT-1y(+)L-type amino acid transporter 1 | |
| SLC7A7 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title