missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC7A7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59856
This item is not returnable.
View return policy
Description
SLC7A7 Polyclonal specifically detects SLC7A7 in Human samples. It is validated for Western Blot.
Specifications
| SLC7A7 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| LAT3, LPI, Monocyte amino acid permease 2, MOP-2, solute carrier family 7 (cationic amino acid transporter, y+ system), member 7, Solute carrier family 7 member 7, Y+L amino acid transporter 1, Y+LAT1, y+LAT-1y(+)L-type amino acid transporter 1 | |
| Rabbit | |
| 56 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Guinea pig 79%, Mouse 79%, Rat 79%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9UM01 | |
| SLC7A7 | |
| Synthetic peptides corresponding to SLC7A7(solute carrier family 7 (cationic amino acid transporter, y+ system), member 7) The peptide sequence was selected from the middle region of SLC7A7. Peptide sequence WGTLVQDIFTYAKVLALIAVIVAGIVRLGQGASTHFENSFEGSSFAVGDI The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 9056 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction