missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC7A4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | SLC7A4 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SLC7A4 Polyclonal specifically detects SLC7A4 in Human samples. It is validated for Western Blot.Specifications
| SLC7A4 | |
| Polyclonal | |
| Rabbit | |
| CAT4cationic amino acid transporter 4, CAT-4MGC129977, HCAT3, Ig heavy chain variable region, MGC129976, solute carrier family 7 (cationic amino acid transporter, y+ system), member 4, Solute carrier family 7 member 4, VH, VH 3 family | |
| SLC7A4 | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 6545 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title