missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC5A5/Sodium Iodide Symporter Antibody (14F), Novus Biologicals™
Mouse Monoclonal Antibody
£176.00 - £427.00
Specifications
| Antigen | SLC5A5/Sodium Iodide Symporter |
|---|---|
| Clone | 14F |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18222935
|
Novus Biologicals
NBP2-59341 |
100 μg |
£427.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608928
|
Novus Biologicals
NBP2-59341-25ug |
25 μg |
£176.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
SLC5A5/Sodium Iodide Symporter Monoclonal specifically detects SLC5A5/Sodium Iodide Symporter in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Microarray.Spezifikation
| SLC5A5/Sodium Iodide Symporter | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1 | |
| SLC5A5 | |
| IgG1 | |
| Protein G purified |
| 14F | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 6528 | |
| Mannose binding protein hNIS fusion (AA468-643) | |
| Primary | |
| Detects 97kDa, non-glycosylated version at 68kDa. Other minor bands associated with hNIS at 160kDa, and degradation products at 30 kDa, and 15kDa. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts