missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC5A5/Sodium Iodide Symporter Antibody (14F), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59341
This item is not returnable.
View return policy
Description
SLC5A5/Sodium Iodide Symporter Monoclonal specifically detects SLC5A5/Sodium Iodide Symporter in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Microarray.
Specifications
| SLC5A5/Sodium Iodide Symporter | |
| Monoclonal | |
| Unconjugated | |
| PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
| SLC5A5 | |
| Mannose binding protein hNIS fusion (AA468-643) | |
| 100 μg | |
| Primary | |
| Detects 97kDa, non-glycosylated version at 68kDa. Other minor bands associated with hNIS at 160kDa, and degradation products at 30 kDa, and 15kDa. | |
| Store at -20C. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 14F | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Na(+)/I(-) cotransporter, Na(+)/I(-) symporter, NISNa(+)/I(-)-symporter, sodium/iodide cotransporter, Sodium-iodide symporter, solute carrier family 5 (sodium iodide symporter), member 5, Solute carrier family 5 member 5, TDH1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6528 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction