missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC45A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£388.00
Specifications
| Antigen | SLC45A2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SLC45A2 Polyclonal specifically detects SLC45A2 in Human samples. It is validated for Western Blot.Specifications
| SLC45A2 | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer | |
| AIM-1, AIM11A1, MATPmembrane associated transporter, Melanoma antigen AIM1, membrane-associated transporter protein, Protein AIM-1, SHEP5, Solute carrier family 45 member 2, solute carrier family 45, member 2, underwhite | |
| SLC45A2 | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q6P2P0 | |
| 51151 | |
| IgG |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto