missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC44A4 Biosimilar Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
£397.00 - £1002.00
Specifications
| Antigen | SLC44A4 Biosimilar |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30283845
![]() |
Invitrogen™
MA559001 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30283555
|
Invitrogen™
MA559002 |
1 mg |
£1002.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term ASG-5ME.
SLC44A4 (solute carrier family 44 member A4) is a gene encoding a protein that primarily functions as the thiamine pyrophosphate transporter (TPPT) in humans. This protein is responsible for the absorption of thiamine pyrophosphate (TPP), a major form of vitamin B1, generated by gut microbiota in the large intestine. The SLC44A4 gene is highly expressed in the colon and is localized to the apical membrane of colonic cells, enabling efficient uptake of TPP. This transporter operates in a sodium-independent manner and is crucial for maintaining thiamine homeostasis, which is vital for various cellular processes including carbohydrate metabolism and neural function. Disruption or mutations in SLC44A4 can impact the body's ability to absorb thiamine, leading to deficiencies that may result in metabolic and neurological disorders.Specifications
| SLC44A4 Biosimilar | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG2SA | |
| Human | |
| Primary | |
| Protein A |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title