Learn More
Invitrogen™ SLC44A4 Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559001
 View more versions of this product
Product Code. 30283845
  • £397.00 / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term ASG-5ME.
SLC44A4 (solute carrier family 44 member A4) is a gene encoding a protein that primarily functions as the thiamine pyrophosphate transporter (TPPT) in humans. This protein is responsible for the absorption of thiamine pyrophosphate (TPP), a major form of vitamin B1, generated by gut microbiota in the large intestine. The SLC44A4 gene is highly expressed in the colon and is localized to the apical membrane of colonic cells, enabling efficient uptake of TPP. This transporter operates in a sodium-independent manner and is crucial for maintaining thiamine homeostasis, which is vital for various cellular processes including carbohydrate metabolism and neural function. Disruption or mutations in SLC44A4 can impact the body's ability to absorb thiamine, leading to deficiencies that may result in metabolic and neurological disorders.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC44A4 Biosimilar
Recombinant Monoclonal
Unconjugated
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A
RUO
Human
Antibody
IgG2SA