missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-62528
This item is not returnable.
View return policy
Description
SLC39A12 Polyclonal specifically detects SLC39A12 in Human samples. It is validated for Western Blot.
Specifications
| SLC39A12 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| bA570F3.1, FLJ30499, LIV-1 subfamily of ZIP zinc transporter 8, LZT-Hs8, MGC43205, MGC51099, solute carrier family 39 (metal ion transporter), member 12, solute carrier family 39 (zinc transporter), member 12, Solute carrier family 39 member 12, zinc transporter ZIP12, ZIP12, ZIP-12, Zrt- and Irt-like protein 12 | |
| Rabbit | |
| 73 kDa | |
| 100 μL | |
| Primary | |
| Equine: 85%; Bovine: 77%. | |
| Human, Bovine, Equine | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q504Y0-3 | |
| SLC39A12 | |
| Synthetic peptides corresponding to SLC39A12(solute carrier family 39 (zinc transporter), member 12) The peptide sequence was selected from the N terminal of SLC39A12. Peptide sequence MCFRTKLSVSWVPLFLLLSRVFSTETDKPSAQDSRSRGSSGQPADLLQVL. | |
| Affinity purified | |
| RUO | |
| 221074 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction