missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£122.57 - £303.85
Specifications
| Antigen | SLC38A1 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Amino acid transporters play essential roles in the uptake of nutrients, production of energy, chemical metabolism, detoxification, and neurotransmitter cycling. SLC38A1 is an important transporter of glutamine, an intermediate in the detoxification of ammonia and the production of urea. Glutamine serves as a precursor for the synaptic transmitter, glutamate.Specifications
| SLC38A1 | |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| Q8K2P7, Q9H2H9, Q9JM15 | |
| 105727, 170567, 81539 | |
| SLC38A1 Fusion Protein Ag2668 | |
| Primary | |
| -20°C | |
| SLC38A1 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Amino acid transporter A1, ATA1, NAT2, SAT1, SLC38A1, SNAT1 | |
| Slc38a1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title