missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12039-1-AP-150UL
This item is not returnable.
View return policy
Description
Amino acid transporters play essential roles in the uptake of nutrients, production of energy, chemical metabolism, detoxification, and neurotransmitter cycling. SLC38A1 is an important transporter of glutamine, an intermediate in the detoxification of ammonia and the production of urea. Glutamine serves as a precursor for the synaptic transmitter, glutamate.Specifications
| SLC38A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC38A1 | |
| Amino acid transporter A1, ATA1, NAT2, SAT1, SLC38A1, SNAT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 105727, 170567, 81539 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8K2P7, Q9H2H9, Q9JM15 | |
| Slc38a1 | |
| SLC38A1 Fusion Protein Ag2668 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction