missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Slc30A8 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

£155.00 - £432.00

Specifications

Antigen Slc30A8
Concentration 0.5 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17710834
GREENER_CHOICE
View Documents
Invitrogen™
PA5119426
200 μL
£432.00
200µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17713004
View Documents
Invitrogen™
PA5121603
20 μL
£155.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Slc30A8 Polyclonal Antibody for Western Blot, IHC (P)

Slc30A8 is a zinc efflux transporter involved in the accumulation of zinc in intracellular vesicles and is expressed at a high level in the pancreas, particularly in islets of Langerhans, and co-localizes with insulin in the secretory pathway granules of the insulin-secreting INS-1 cells. Allelic variants of Slc30A8 have been identified as major genetic risk factors for the development of Type 2 diabetes, but no genetic association has been found with Type 1 diabetes. Slc30A8 expression in b-cells was found to be influenced by cytokine expression, particularly IFN-gamma and IL-1 beta.
Specifications

Specifications

Slc30A8
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human, Mouse
SLC30A8
C820002P14Rik; LOW QUALITY PROTEIN: uncharacterized protein slc30a8; Slc30a8; solute carrier family 30 (zinc transporter), member 8; solute carrier family 30 member 8; wu:fe49g03; Zinc transporter 8; zinc transporter 8; LOW QUALITY PROTEIN: zinc transporter 8; zinc transporter ZnT-8; Znt8; ZnT-8
SLC30A8
Primary
Antigen affinity chromatography, Protein A
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q8BGG0, Q8IWU4
169026, 239436
Recombinant protein Solute Carrier Family 30 Member 8. The antigen corresponds to amino acid range 263-369 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.