missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Slc30A8 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5121603

 View more versions of this product

Product Code. 17713004

  • £155.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Slc30A8 Polyclonal Antibody for Western Blot, IHC (P)

Slc30A8 is a zinc efflux transporter involved in the accumulation of zinc in intracellular vesicles and is expressed at a high level in the pancreas, particularly in islets of Langerhans, and co-localizes with insulin in the secretory pathway granules of the insulin-secreting INS-1 cells. Allelic variants of Slc30A8 have been identified as major genetic risk factors for the development of Type 2 diabetes, but no genetic association has been found with Type 1 diabetes. Slc30A8 expression in b-cells was found to be influenced by cytokine expression, particularly IFN-gamma and IL-1 beta.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Slc30A8
Polyclonal
Unconjugated
SLC30A8
C820002P14Rik; LOW QUALITY PROTEIN: uncharacterized protein slc30a8; Slc30a8; solute carrier family 30 (zinc transporter), member 8; solute carrier family 30 member 8; wu:fe49g03; Zinc transporter 8; zinc transporter 8; LOW QUALITY PROTEIN: zinc transporter 8; zinc transporter ZnT-8; Znt8; ZnT-8
Rabbit
Antigen affinity chromatography, Protein A
RUO
169026, 239436
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q8BGG0, Q8IWU4
SLC30A8
Recombinant protein Solute Carrier Family 30 Member 8. The antigen corresponds to amino acid range 263-369 of the target protein.
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.