missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC30A4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94290-0.1ml
This item is not returnable.
View return policy
Description
SLC30A4 Polyclonal antibody specifically detects SLC30A4 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SLC30A4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| solute carrier family 30 (zinc transporter), member 4, zinc transporter 4, znT-4, ZNT4Solute carrier family 30 member 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 335-429 of human SLC30A4 (NP_037441.2). VPSHLNVDYIKEALMKIEDVYSVEDLNIWSLTSGKSTAIVHIQLIPGSSSKWEEVQSKANHLLLNTFGMYRCTIQLQSYRQEVDRTCANCQSSSP | |
| 0.1 mL | |
| Endocrinology, Neuroscience, Signal Transduction | |
| 7782 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction