missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC2A5 Monoclonal Antibody (14C8)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA1036X
This item is not returnable.
View return policy
Description
This antibody requires overnight incubation in a Western blot for optimal performance. Do not use this antibody in a Fast Western blotting procedure.
Cytochalasin B-sensitive carrier. Seems to function primarily as a fructose transporter.
Specifications
| SLC2A5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1 mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
| P22732, Q9WV38 | |
| SLC2A5 | |
| Protein expressed in 293T cell transfected with human SLC2A5 expression vector. | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 14C8 | |
| Unconjugated | |
| SLC2A5 | |
| AI526984; Fructose transporter; glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT5; GLUT-5; RP5-963K15.1; SLC2A5; Slc5a; solute carrier family 2 (facilitated glucose transporter), member 5; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; solute carrier family 2 member 5; solute carrier family 2, facilitated glucose transporter member 5; solute carrier family 2, member 5; testicular tissue protein Li 81 | |
| Mouse | |
| Protein A | |
| RUO | |
| 56485, 6518 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction