missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC2A5 Monoclonal Antibody (14C8)

Mouse Monoclonal Antibody

£188.00 - £477.00

Specifications

Antigen SLC2A5
Clone 14C8
Concentration 1 mg/mL
Content And Storage -20°C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12867762
View Documents
Invitrogen™
MA1036
100 μg
£477.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12877762
View Documents
Invitrogen™
MA1036X
20 μL
£188.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody requires overnight incubation in a Western blot for optimal performance. Do not use this antibody in a Fast Western blotting procedure.

Cytochalasin B-sensitive carrier. Seems to function primarily as a fructose transporter.
Specifications

Specifications

SLC2A5
1 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Mouse
RUO
PBS with 1 mg/mL BSA, 30% glycerol and 0.05% sodium azide
P22732, Q9WV38
56485, 6518
Protein expressed in 293T cell transfected with human SLC2A5 expression vector.
Antibody
14C8
-20°C
Monoclonal
Liquid
IgG1
Human, Mouse
SLC2A5
AI526984; Fructose transporter; glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT5; GLUT-5; RP5-963K15.1; SLC2A5; Slc5a; solute carrier family 2 (facilitated glucose transporter), member 5; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; solute carrier family 2 member 5; solute carrier family 2, facilitated glucose transporter member 5; solute carrier family 2, member 5; testicular tissue protein Li 81
SLC2A5
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.