missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | SLC22A9 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SLC22A9 Polyclonal antibody specifically detects SLC22A9 in Human, Mouse samples. It is validated for Western BlotSpecifications
| SLC22A9 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| FLJ23666, HOAT4, OAT4UST3H, OAT7, Organic anion transporter 4, organic anion transporter 7, solute carrier family 22 (organic anion transporter), member 9, solute carrier family 22 (organic anion/cation transporter), member 9, solute carrier family 22 member 9, UST3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-145 of human SLC22A9 (NP_543142.2). FTAFIPGHRCWVHILDNDTVSDNDTGALSQDALLRISIPLDSNMRPEKCRRFVHPQWQLLHLNGTFPNTSDADMEPCVDGWVYDRISFSSTIVTEWDLVCDSQSLT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 114571 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title