missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | SLC22A3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230328
|
Novus Biologicals
NBP3-35881-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227081
|
Novus Biologicals
NBP3-35881-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC22A3 Polyclonal antibody specifically detects SLC22A3 in Mouse samples. It is validated for ELISA,Western BlotSpecifications
| SLC22A3 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| EMTEMTH, Extraneuronal monoamine transporter, OCT3EMT organic cation transporter 3, Organic cation transporter 3, solute carrier family 22 (extraneuronal monoamine transporter), member 3, solute carrier family 22 member 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 289-349 of human SLC22A3 (NP_068812.1).,, Sequence:, PESPRWLITRKKGDKALQILRRIAKCNGKYLSSNYSEITVTDEEVSNPSFLDLVRTPQMRK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 6581 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title